missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpiperidine, 98%

Catalog No. AC161930050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161930050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC161930050 Supplier Thermo Scientific Chemicals Supplier No. 161930050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 31252-42-3 | C12H17N | 175.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31252-42-3
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.28
MDL Number MFCD00006006
InChI Key ABGXADJDTPFFSZ-UHFFFAOYSA-N
Synonym piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
PubChem CID 31738
IUPAC Name 4-benzylpiperidine
SMILES C(C1CCNCC1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 6.0°C to 7.0°C
Color Colorless to Yellow
Density 0.9900g/mL
Boiling Point 279.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5370 to 1.5390
Quantity 5 g
Beilstein 20, 296
Specific Gravity 0.99
Formula Weight 175.27
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 4-Benzylpiperidine, 98%
Show More Show Less
EINECSNumber 250-535-0
RTECSNumber TM4728000
TSCA TSCA

RUO – Research Use Only