missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzylphenol, Thermo Scientific™
Specifications
Specifications
| Alpha Vector | BENZYLPHENOL |
| Melting Point | 80°C to 85°C |
| Color | White to beige to brown |
| Infrared Spectrum | Conforms |
| Packaging | Glass Bottle |
| Linear Formula | C6H5CH2C6H4OH |
| Quantity | 1 g |
| Beilstein | 06-675 |
| Merck Index | 15-1145 |
| Physical Form | Crystalline Powder or Crystals |
| Show More |
Safety and Handling
| Hazard Category | Exclamation mark |
| EINECSNumber | 202-950-3 |
| RTECSNumber | SM8330500 |
| Recommended Storage | Normal conditions |