missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzyloxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$57.79 - $57.79
Chemical Identifiers
| CAS | 146631-00-7 |
|---|---|
| Molecular Formula | C13H13BO3 |
| Molecular Weight (g/mol) | 228.054 |
| MDL Number | MFCD01075705 |
| InChI Key | DMJHEIDWSIAXCS-UHFFFAOYSA-N |
| PubChem CID | 2734314 |
| IUPAC Name | (4-phenylmethoxyphenyl)boronic acid |
| SMILES | B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O |
Chemical Identifiers
| 146631-00-7 | |
| 228.054 | |
| DMJHEIDWSIAXCS-UHFFFAOYSA-N | |
| (4-phenylmethoxyphenyl)boronic acid |
| C13H13BO3 | |
| MFCD01075705 | |
| 2734314 | |
| B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O |
Specifications
| 146631-00-7 | |
| MFCD01075705 | |
| 4-benzyloxyphenylboronic acid, 4-benzyloxybenzeneboronic acid, 4-benzyloxy phenyl boronic acid, 4-benzyloxy phenylboronic acid, 4-benzyloxyphenyl boronic acid, 4-benzyl-oxy phenyl boranediol, boronic acid, 4-phenylmethoxy phenyl, 4-phenylmethoxyphenyl boronic acid, pubchem9561 | |
| B(C1=CC=C(C=C1)OCC2=CC=CC=C2)(O)O | |
| 228.054 | |
| 228.05 | |
| 4-Benzyloxyphenylboronic Acid (contains varying amounts of Anhydride) |
| C13H13BO3 | |
| 1 g | |
| DMJHEIDWSIAXCS-UHFFFAOYSA-N | |
| (4-phenylmethoxyphenyl)boronic acid | |
| 2734314 | |
| Crystalline Powder |
Safety and Handling
TSCA : No