missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. p-7193745
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47251G Supplier TCI America Supplier No. B47251G

Chemical Identifiers

CAS 184000-11-1
Molecular Formula C14H13BO4
Molecular Weight (g/mol) 256.06
MDL Number MFCD02179442
InChI Key UXSDJFGNRVVDFM-UHFFFAOYSA-N
PubChem CID 2773251
IUPAC Name {4-[(benzyloxy)carbonyl]phenyl}boronic acid
SMILES OB(O)C1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 1 g
Synonym 4-benzyloxycarbonylphenylboronic acid, 4-benzyloxycarbonylphenyl boronic acid, 4-benzyloxycarbonyl benzeneboronic acid, 4-benzyloxy carbonyl phenyl boronic acid, 4-benzyloxycarbonyl phenylboronic acid, p-benzyloxycarbonylphenylboronic acid, benzyl 4-boronobenzoate, 4-benzyloxy carbonyl phenylboronic acid, 4-phenylmethoxycarbonylphenyl boronic acid
Formula Weight 256.06
Physical Form Crystalline Powder
Chemical Name or Material 4-(Benzyloxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)