missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxybromobenzene, 97%, Thermo Scientific™

Catalog No. p-4813063
Change view
Click to view available options
Quantity:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442460100 10g
AC442460500 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442460100 Supplier Thermo Scientific Chemicals Supplier No. 442460100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 6793-92-6
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.14
InChI Key OUQSGILAXUXMGI-UHFFFAOYSA-N
Synonym 4-benzyloxybromobenzene, 1-benzyloxy-4-bromobenzene, 1-bromo-4-phenylmethoxy benzene, 1-bromo-4-benzyloxybenzene, 4-benzyloxy bromobenzene, 1-benzyloxy-4-bromo-benzene, 1-bromo-4-phenylmethyl oxy benzene, 4-benzyloxy-bromobenzene, p-benzyloxy bromobenzene
PubChem CID 138835
IUPAC Name 1-bromo-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)Br

Specifications

Boiling Point 166°C to 167°C (4.0 Torr)
Assay Percent Range 96% min. (GC)
Quantity 10g
Formula Weight 263.14
Percent Purity 97%
Chemical Name or Material 4-Benzyloxybromobenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

RUO – Research Use Only