missing translation for 'onlineSavingsMsg'
Learn More

4'-Benzyloxyacetophenone 98.0+%, TCI America™

Catalog No. B33655G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33655G 5 g
B33651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33655G Supplier TCI America Supplier No. B33655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54696-05-8
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00017247
InChI Key MKYMYZJJFMPDOA-UHFFFAOYSA-N
Synonym 4'-benzyloxyacetophenone, 1-4-benzyloxy phenyl ethanone, 4-benzyloxyacetophenone, 4'-benzyloxy-acetophenone, 1-4-benzyloxy phenyl ethan-1-one, ethanone, 1-4-phenylmethoxy phenyl, 4'-benzyloxy acetophenone, 1-4-benzyloxyphenyl ethanone
PubChem CID 245226
IUPAC Name 1-(4-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)OCC2=CC=CC=C2

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 5 g
Formula Weight 226.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Benzyloxyacetophenone