missing translation for 'onlineSavingsMsg'
Learn More

4'-Benzyloxyacetophenone, 98%

Catalog No. p-7024100
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1830206 5 g
AAA1830214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1830206 Supplier Thermo Scientific Chemicals Supplier No. A1830206
Only null left

CAS: 54696-05-8 | C15H14O2 | 226.275 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54696-05-8
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00017247
InChI Key MKYMYZJJFMPDOA-UHFFFAOYSA-N
Synonym 4'-benzyloxyacetophenone, 1-4-benzyloxy phenyl ethanone, 4-benzyloxyacetophenone, 4'-benzyloxy-acetophenone, 1-4-benzyloxy phenyl ethan-1-one, ethanone, 1-4-phenylmethoxy phenyl, 4'-benzyloxy acetophenone, 1-4-benzyloxyphenyl ethanone
PubChem CID 245226
IUPAC Name 1-(4-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)OCC2=CC=CC=C2

Specifications

Melting Point 91°C to 92°C
Quantity 5 g
Beilstein 1912124
Formula Weight 226.28
Percent Purity 98%
Chemical Name or Material 4'-Benzyloxyacetophenone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only