missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-6-chloropyrimidine, 95%

Catalog No. aah2739003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2739003 1 g
AAH27390MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2739003 Supplier Thermo Scientific Chemicals Supplier No. H2739003
Only null left

CAS: 405930-65-6 | C11H9ClN2O | 220.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 405930-65-6
Molecular Formula C11H9ClN2O
Molecular Weight (g/mol) 220.66
MDL Number MFCD06798234
InChI Key KXDXFVQZZGSEBU-UHFFFAOYSA-N
Synonym 4-benzyloxy-6-chloropyrimidine, 6-benzyloxy-4-chloropyrimidine, pyrimidine, 4-chloro-6-phenylmethoxy, acmc-20amoq, 4-chloro-6-benzyloxypyrimidine, pyrimidine,4-chloro-6-phenylmethoxy
PubChem CID 22599871
IUPAC Name 4-chloro-6-phenylmethoxypyrimidine
SMILES ClC1=NC=NC(OCC2=CC=CC=C2)=C1

Specifications

Quantity 1 g
Formula Weight 220.66
Percent Purity 95%
Chemical Name or Material 4-Benzyloxy-6-chloropyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only