missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzyloxy-5-bromo-2-chloropyrimidine, 97%, Thermo Scientific™
$255.97 - $255.97
Chemical Identifiers
| CAS | 205672-19-1 |
|---|---|
| Molecular Formula | C11H8BrClN2O |
| Molecular Weight (g/mol) | 299.552 |
| MDL Number | MFCD06798237 |
| InChI Key | PFOACTPILUWRCT-UHFFFAOYSA-N |
| Synonym | 4-benzyloxy-5-bromo-2-chloropyrimidine, 5-bromo-2-chloro-4-phenylmethoxy pyrimidine, pyrimidine, 5-bromo-2-chloro-4-phenylmethoxy, pubchem6890, acmc-20aig5, 4-benzyloxy-5-bromo-2-chloro-pyrimidine, 2-chloro-4-benzyloxy-5-bromopyrimidine, 5-bromanyl-2-chloranyl-4-phenylmethoxy-pyrimidine |
| PubChem CID | 10732937 |
| IUPAC Name | 5-bromo-2-chloro-4-phenylmethoxypyrimidine |
| SMILES | C1=CC=C(C=C1)COC2=NC(=NC=C2Br)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2770603
|
Thermo Scientific Chemicals
H2770603 |
1 g |
Each for $255.97
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 205672-19-1 | |
| 299.552 | |
| PFOACTPILUWRCT-UHFFFAOYSA-N | |
| 10732937 | |
| C1=CC=C(C=C1)COC2=NC(=NC=C2Br)Cl |
| C11H8BrClN2O | |
| MFCD06798237 | |
| 4-benzyloxy-5-bromo-2-chloropyrimidine, 5-bromo-2-chloro-4-phenylmethoxy pyrimidine, pyrimidine, 5-bromo-2-chloro-4-phenylmethoxy, pubchem6890, acmc-20aig5, 4-benzyloxy-5-bromo-2-chloro-pyrimidine, 2-chloro-4-benzyloxy-5-bromopyrimidine, 5-bromanyl-2-chloranyl-4-phenylmethoxy-pyrimidine | |
| 5-bromo-2-chloro-4-phenylmethoxypyrimidine |
Specifications
| 205672-19-1 | |
| C11H8BrClN2O | |
| 1 g | |
| PFOACTPILUWRCT-UHFFFAOYSA-N | |
| 5-bromo-2-chloro-4-phenylmethoxypyrimidine | |
| 10732937 | |
| 97% |
| 114°C to 116°C | |
| MFCD06798237 | |
| 4-benzyloxy-5-bromo-2-chloropyrimidine, 5-bromo-2-chloro-4-phenylmethoxy pyrimidine, pyrimidine, 5-bromo-2-chloro-4-phenylmethoxy, pubchem6890, acmc-20aig5, 4-benzyloxy-5-bromo-2-chloro-pyrimidine, 2-chloro-4-benzyloxy-5-bromopyrimidine, 5-bromanyl-2-chloranyl-4-phenylmethoxy-pyrimidine | |
| C1=CC=C(C=C1)COC2=NC(=NC=C2Br)Cl | |
| 299.552 | |
| 299.56 | |
| 4-Benzyloxy-5-bromo-2-chloropyrimidine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures