missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-5-bromo-2-chloropyrimidine, 97%, Thermo Scientific™

Catalog No. AAH2770603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2770603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2770603 Supplier Thermo Scientific Chemicals Supplier No. H2770603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 205672-19-1
Molecular Formula C11H8BrClN2O
Molecular Weight (g/mol) 299.552
MDL Number MFCD06798237
InChI Key PFOACTPILUWRCT-UHFFFAOYSA-N
Synonym 4-benzyloxy-5-bromo-2-chloropyrimidine, 5-bromo-2-chloro-4-phenylmethoxy pyrimidine, pyrimidine, 5-bromo-2-chloro-4-phenylmethoxy, pubchem6890, acmc-20aig5, 4-benzyloxy-5-bromo-2-chloro-pyrimidine, 2-chloro-4-benzyloxy-5-bromopyrimidine, 5-bromanyl-2-chloranyl-4-phenylmethoxy-pyrimidine
PubChem CID 10732937
IUPAC Name 5-bromo-2-chloro-4-phenylmethoxypyrimidine
SMILES C1=CC=C(C=C1)COC2=NC(=NC=C2Br)Cl

Specifications

Melting Point 114°C to 116°C
Quantity 1 g
Formula Weight 299.56
Percent Purity 97%
Chemical Name or Material 4-Benzyloxy-5-bromo-2-chloropyrimidine
TSCA No
Recommended Storage Ambient temperatures