missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methylbenzeneboronic acid, 96%

Catalog No. p-7035356
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5307303 1 g
AAH5307306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5307303 Supplier Thermo Scientific Chemicals Supplier No. H5307303

CAS: 338454-30-1 | C14H15BO3 | 242.081 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 338454-30-1
Molecular Formula C14H15BO3
Molecular Weight (g/mol) 242.081
MDL Number MFCD06409944
InChI Key QPXFEWQLALNXEZ-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methylphenylboronic acid, 4-benzyloxy-3-methylphenyl boronic acid, 4-benzyloxy-3-methylbenzeneboronic acid, acmc-1agrw, 4-benzyloxy-3-methylphenylboronicacid, 4-benzyloxy-3-methyl-phenylboron acid, 4-benzyloxy-3-methylphenyl boronicacid
PubChem CID 17750522
IUPAC Name (3-methyl-4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)C)(O)O

Specifications

Melting Point 102°C to 106°C
Quantity 1 g
Formula Weight 242.08
Percent Purity 96%
Chemical Name or Material 4-Benzyloxy-3-methylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only