missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2-methylbenzeneboronic acid, 98%

Catalog No. p-7035548
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5326603 1 g
AAH5326606 5 g
2 options

Catalog No. AAH5326603

Supplier: Thermo Scientific Chemicals H5326603

Only null left

CAS: 847560-49-0 | C14H15BO3 | 242.081 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 847560-49-0
Molecular Formula C14H15BO3
Molecular Weight (g/mol) 242.081
MDL Number MFCD03788404
InChI Key VCDFDGOVNBMYRW-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-methylphenylboronic acid, 4-benzyloxy-2-methylphenyl boronic acid, 4-benzyloxy-2-methylbenzeneboronic acid, 5-benzyloxytoluene-2-boronic acid, acmc-209pwd, 2-methyl-4-benzyloxyphenylboronic acid, 4-benzyloxy-2-methylphenyl boronicacid
PubChem CID 2773245
IUPAC Name (2-methyl-4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)C)(O)O

Specifications

Melting Point 154°C to 158°C
Quantity 1 g
Formula Weight 242.08
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-2-methylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only