missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2-methylbenzaldehyde 97.0+%, TCI America™

Catalog No. b5136200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5136200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5136200MG Supplier TCI America Supplier No. B5136200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101093-56-5
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD01317809
InChI Key OGVPJBJWJCZBTH-UHFFFAOYSA-N
Synonym 4-Benzyloxy-o-tolualdehyde
PubChem CID 2759806
IUPAC Name 2-methyl-4-phenylmethoxybenzaldehyde
SMILES CC1=C(C=CC(=C1)OCC2=CC=CC=C2)C=O

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 234°C
Quantity 200 mg
Formula Weight 226.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyloxy-2-methylbenzaldehyde