missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2-hydroxypyridine, 97%

Catalog No. AAB2477406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2477406 5 g
AAB2477414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2477406 Supplier Thermo Scientific Chemicals Supplier No. B2477406

CAS: 53937-02-3 | C12H11NO2 | 201.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53937-02-3
Molecular Formula C12H11NO2
Molecular Weight (g/mol) 201.23
MDL Number MFCD04039771
InChI Key DOVNUEPFPBWTSV-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one
PubChem CID 3607724
SMILES O=C1NC=CC(OCC2=CC=CC=C2)=C1

Specifications

Melting Point 194°C to 198°C
Quantity 5 g
IUPAC Name 4-(benzyloxy)-1,2-dihydropyridin-2-one
Formula Weight 201.22
Percent Purity 97%
Chemical Name or Material 4-Benzyloxy-2-hydroxypyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only