missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl N-(tert-Butoxycarbonyl)-L-aspartate 98.0+%, TCI America™

Catalog No. p-7119169
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16285G 5 g
B162825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16285G Supplier TCI America Supplier No. B16285G
  • Purity (Neutralization titration): ≥98.0%
  • Molecular Formula: C16H21NO6
  • Molecular Weight: 323.35 g/mol
  • Physical State: Solid (White to Almost white powder to crystal)
  • Melting Point: 102°C to 104°C
  • Specific Rotation [α]20/D: -19.0 to -21.0 deg (C=2, DMF)
  • Solubility: Almost transparent in N,N-DMF
  • NMR: Confirmed to structure

Chemical Identifiers

CAS 7536-58-5
Molecular Formula C16H21NO6
Molecular Weight (g/mol) 323.345
MDL Number MFCD00065564
InChI Key SOHLZANWVLCPHK-LBPRGKRZSA-N
Synonym boc-asp obzl-oh, boc-l-aspartic acid 4-benzyl ester, s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-l-asp obzl-oh, boc-l-aspartic acid-4-benzyl ester, 4-benzyl n-boc-l-aspartate, boc-l-aspartic acid 4-benzylester, 2s-4-benzyloxy-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, boc-aspartic acid beta-benzyl ester, n-tert-butoxycarbonyl-l-aspartic acid 4-benzyl ester
PubChem CID 1581888
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)OCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 103°C
Color White
Quantity 5 g
Formula Weight 323.35
Percent Purity ≥98.0% (T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyl N-(tert-Butoxycarbonyl)-L-aspartate