missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl N-Carbobenzoxy-L-aspartate 98.0+%, TCI America™

Catalog No. p-7123235
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39035G 5 g
B390325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39035G Supplier TCI America Supplier No. B39035G
Only null left

Chemical Identifiers

CAS 3479-47-8
Molecular Formula C19H19NO6
Molecular Weight (g/mol) 357.36
MDL Number MFCD00037820
InChI Key VUKCNAATVIWRTF-UHFFFAOYNA-N
Synonym z-asp obzl-oh, cbz-asp obzl-oh, n-cbz-l-aspartic acid 4-benzyl ester, z-l-aspartic acid 4-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-benzyl ester, 4-benzyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid, n-carbobenzoxy-l-aspartic acid 4-benzyl ester, 2s-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid, 2s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid
PubChem CID 47602
IUPAC Name 4-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid
SMILES OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 110°C
Color White
Quantity 5 g
Formula Weight 357.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyl N-Carbobenzoxy-L-aspartate