missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-4-hydroxypiperidine, 97%

Catalog No. aaa1566414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1566414 25 g
AAA1566403 1 g
AAA1566406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1566414 Supplier Thermo Scientific Chemicals Supplier No. A1566414
Only null left

CAS: 51135-96-7 | C12H17NO | 191.274 g/mol

It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a building block used for the synthesis of various compounds, having therapeutic properties, such as synthesis of 4-Benzimidazolyl-piperidinylcarbonyl-piperidine analogs as histamine H3 antagonists.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 51135-96-7
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00023147
InChI Key KJZBZOFESQSBCV-UHFFFAOYSA-N
Synonym 4-benzyl-4-hydroxypiperidine, 4-piperidinol, 4-benzyl, 4-benzyl-4-piperidinol, 4-piperidinol, 4-phenylmethyl, acmc-20amoy, pubchem16262, 4benzyl-piperidin-4-ol, maybridge3_002978, 4-benzyl-piperidin-4-ol, 4-benzyl-4-piperidinol #
PubChem CID 98152
IUPAC Name 4-benzylpiperidin-4-ol
SMILES C1CNCCC1(CC2=CC=CC=C2)O

Specifications

Melting Point ∼80°C
Quantity 25 g
Beilstein 1451803
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material 4-Benzyl-4-hydroxypiperidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 257-003-7
RTECSNumber TN6836000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only