missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione 98.0+%, TCI America™

Catalog No. B44365MG
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44365MG 5 mg
B443625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44365MG Supplier TCI America Supplier No. B44365MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 327036-89-5
Molecular Formula C10H10N2O2S
Molecular Weight (g/mol) 222.26
MDL Number MFCD04973552
InChI Key JDSJDASOXWCHPN-UHFFFAOYSA-N
Synonym TDZD-8
PubChem CID 4124851
IUPAC Name 4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
SMILES CN1SC(=O)N(CC2=CC=CC=C2)C1=O

Specifications

Melting Point 63°C
Color White
Quantity 5 mg
Formula Weight 222.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
TSCA No
Recommended Storage Freezer (-20 °C)