missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-2-hydroxymorpholin-3-one 98.0+%, TCI America™

Catalog No. b49185g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49185G 5 g
B49181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49185G Supplier TCI America Supplier No. B49185G

Chemical Identifiers

CAS 287930-73-8
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
MDL Number MFCD07776435
InChI Key CAGSEMPCDRYWFN-UHFFFAOYSA-N
PubChem CID 9942497
IUPAC Name 4-benzyl-2-hydroxymorpholin-3-one
SMILES C1COC(C(=O)N1CC2=CC=CC=C2)O

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Formula Weight 207.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzyl-2-hydroxymorpholin-3-one