missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylpyridine 99.0+%, TCI America™

Catalog No. B030625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B030625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B030625G Supplier TCI America Supplier No. B030625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14548-46-0
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006430
InChI Key SKFLCXNDKRUHTA-UHFFFAOYSA-N
Synonym 4-benzoylpyridine, phenyl pyridin-4-yl methanone, phenyl 4-pyridyl ketone, methanone, phenyl-4-pyridinyl, phenyl 4-pyridinyl methanone, 4-pyridyl phenyl ketone, pyridine, 4-benzoyl, ketone, phenyl 4-pyridyl, gamma-benzoylpyridine, 4-benzoyl pyridine
PubChem CID 26731
IUPAC Name phenyl(pyridin-4-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=NC=C2

Specifications

Melting Point 72°C
Color White-Yellow
Boiling Point 130°C
Quantity 25 g
Formula Weight 183.21
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzoylpyridine
RTECSNumber OB6402000
TSCA Yes