missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylpiperidine hydrochloride, 98%

Catalog No. AC232860100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232860100 10 g Glass bottle
AC232860010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC232860100 Supplier Thermo Scientific Chemicals Supplier No. 232860100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25519-80-6 | C12H15NO·ClH | 225.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25519-80-6
Molecular Formula C12H15NO·ClH
Molecular Weight (g/mol) 225.72
MDL Number MFCD00066982
InChI Key NXYKIFZJQXOUJS-UHFFFAOYSA-N
Synonym 4-benzoylpiperidine hydrochloride, phenyl piperidin-4-yl methanone hydrochloride, 4-benzoylpiperidine hcl, phenyl 4-piperidyl methanone hydrochloride, 4-benzoyl piperidine hydrochloride, 4-benzoylpiperidine hydrogen chloride, methanone, phenyl-4-piperidinyl-, hydrochloride, phenyl 4-piperidyl ketone, chloride, 4-benzoyl piperidine hcl, pubchem9881
PubChem CID 2724437
IUPAC Name phenyl(piperidin-4-yl)methanone;hydrochloride
SMILES C1CNCCC1C(=O)C2=CC=CC=C2.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 226°C to 227°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol and ethanol
Formula Weight 225.72
Percent Purity 98%
Chemical Name or Material 4-Benzoylpiperidine hydrochloride, 98%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only