missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbutyric Acid 96.0+%, TCI America™

Catalog No. p-7119754
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B27685G 5 g
B276825G 25 g
2 options

Catalog No. B27685G

Supplier: TCI America B27685G

Only null left

Chemical Identifiers

CAS 1501-05-9
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00004411
InChI Key SHKWSBAVRQZYLE-UHFFFAOYSA-N
Synonym 4-benzoylbutyric acid, 5-oxo-5-phenylvaleric acid, benzenepentanoic acid, .delta.-oxo, 4-benzoyl-butyric acid, timtec-bb sbb007811, 5-oxo-5-phenyl-pentanoic acid, 4-benzoylbutyric acid;4-benzoylbutanoic acid, butanoic acid, 4-benzoyl, 4-benzoylbutyricacid
PubChem CID 73914
IUPAC Name 5-oxo-5-phenylpentanoic acid
SMILES C1=CC=C(C=C1)C(=O)CCCC(=O)O

Specifications

Melting Point 129°C
Quantity 5 g
Formula Weight 192.21
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzoylbutyric Acid