missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbutyric acid, 97%

Catalog No. AC105740250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC105740250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC105740250 Supplier Thermo Scientific Chemicals Supplier No. 105740250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1501-05-9 | C11H12O3 | 192.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 1501-05-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 124°C to 128°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C11H12O3
Linear Formula C6H5CO(CH2)3CO2H
Quantity 25 g
MDL Number MFCD00004411
Beilstein 10, 708
Synonym 4-benzoylbutyric acid, 5-oxo-5-phenylvaleric acid, benzenepentanoic acid, .delta.-oxo, 4-benzoyl-butyric acid, timtec-bb sbb007811, 5-oxo-5-phenyl-pentanoic acid, 4-benzoylbutyric acid;4-benzoylbutanoic acid, butanoic acid, 4-benzoyl, 4-benzoylbutyricacid
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in benzene (warm),chloroform,ethanol,,dichloromethane,ethyl acetate,aqueous alkali,insoluble in hexane
InChI Key SHKWSBAVRQZYLE-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C(=O)CCCC(=O)O
IUPAC Name 5-oxo-5-phenylpentanoic acid
Molecular Weight (g/mol) 192.21
PubChem CID 73914
Formula Weight 192.21
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Benzoylbutyric acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-113-5

RUO – Research Use Only