missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoylbutyric acid, 97%

Catalog No. AAL0401306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0401306 5 g
AAL0401314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0401306 Supplier Thermo Scientific Chemicals Supplier No. L0401306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1501-05-9 | C11H12O3 | 192.214 g/mol

4-Benzoylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzoylbutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1501-05-9
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00004411
InChI Key SHKWSBAVRQZYLE-UHFFFAOYSA-N
Synonym 4-benzoylbutyric acid, 5-oxo-5-phenylvaleric acid, benzenepentanoic acid, .delta.-oxo, 4-benzoyl-butyric acid, timtec-bb sbb007811, 5-oxo-5-phenyl-pentanoic acid, 4-benzoylbutyric acid;4-benzoylbutanoic acid, butanoic acid, 4-benzoyl, 4-benzoylbutyricacid
PubChem CID 73914
IUPAC Name 5-oxo-5-phenylpentanoic acid
SMILES C1=CC=C(C=C1)C(=O)CCCC(=O)O

Specifications

Melting Point 126°C to 128°C
Quantity 5 g
Beilstein 641947
Solubility Information Insoluble in water.
Formula Weight 192.21
Percent Purity 97%
Chemical Name or Material 4-Benzoylbutyric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-113-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only