missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Azidobenzoic Acid 97.0+%, TCI America™
$171.49 - $507.02
Chemical Identifiers
| CAS | 6427-66-3 |
|---|---|
| Molecular Formula | C7H5N3O2 |
| Molecular Weight (g/mol) | 163.136 |
| MDL Number | MFCD00013987 |
| InChI Key | PQXPAFTXDVNANI-UHFFFAOYSA-N |
| PubChem CID | 3034184 |
| IUPAC Name | 4-azidobenzoic acid |
| SMILES | C1=CC(=CC=C1C(=O)O)N=[N+]=[N-] |
Chemical Identifiers
| 6427-66-3 | |
| 163.136 | |
| PQXPAFTXDVNANI-UHFFFAOYSA-N | |
| 4-azidobenzoic acid |
| C7H5N3O2 | |
| MFCD00013987 | |
| 3034184 | |
| C1=CC(=CC=C1C(=O)O)N=[N+]=[N-] |
Specifications
| 6427-66-3 | |
| MFCD00013987 | |
| 1325 | |
| C1=CC(=CC=C1C(=O)O)N=[N+]=[N-] | |
| 163.136 | |
| 163.14 | |
| Crystalline Powder |
| C7H5N3O2 | |
| 5 g | |
| PQXPAFTXDVNANI-UHFFFAOYSA-N | |
| 4-azidobenzoic acid | |
| 3034184 | |
| ≥97.0% (HPLC,T) | |
| 4-Azidobenzoic Acid |
Safety and Handling
EINECSNumber : (3)-1680
TSCA : No