missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Azidobenzoic Acid 97.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| UN Number | 1325 |
| Formula Weight | 163.14 |
| Percent Purity | ≥97.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Azidobenzoic Acid |
Safety and Handling
| EINECSNumber | (3)-1680 |
| TSCA | No |