Learn More
4-Azido-2,3,5,6-tetrafluorobenzoic Acid 98.0+%, TCI America™
Supplier:  TCI America A26741G
Catalog No. A26741G
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
4-Azido-2,3,5,6-tetrafluorobenzoic Acid
139°C
C7HF4N3O2
1 g
IOJFHZXQSLNAQJ-UHFFFAOYSA-N
4-azido-2,3,5,6-tetrafluorobenzoic acid
2762578
≥98.0% (HPLC,T)
122590-77-6
Yellow
MFCD00467436
1325
OC(=O)C1=C(F)C(F)=C(N=[N+]=[N-])C(F)=C1F
235.10
235.10
Crystalline Powder
Chemical Identifiers
122590-77-6
235.10
IOJFHZXQSLNAQJ-UHFFFAOYSA-N
4-azido-2,3,5,6-tetrafluorobenzoic acid
C7HF4N3O2
MFCD00467436
2762578
OC(=O)C1=C(F)C(F)=C(N=[N+]=[N-])C(F)=C1F
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
TSCA : No
Recommended Storage : Freezer