missing translation for 'onlineSavingsMsg'
Learn More

4-Azaxanthone, 97%

Catalog No. AAA1204503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1204503 1 g
AAA1204506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1204503 Supplier Thermo Scientific Chemicals Supplier No. A1204503

CAS: 6537-46-8 | C12H7NO2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6537-46-8
Molecular Formula C12H7NO2
MDL Number MFCD00052253

Specifications

Melting Point 180°C to 182°C
Quantity 1 g
Beilstein 8657
Formula Weight 197.2
Percent Purity 97%
Chemical Name or Material 1-Azaxanthone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only