missing translation for 'onlineSavingsMsg'
Learn More

4-Amylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7117635
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
A19265G 5 g
A192625G 25 g
2 options

Catalog No. A19265G

Supplier: TCI America A19265G

Only null left

Chemical Identifiers

CAS 54410-90-1
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
InChI Key VHWGPISIUNUREA-UHFFFAOYSA-N
Synonym 4-Pentylcyclohexanol
PubChem CID 41076
IUPAC Name 4-pentylcyclohexan-1-ol
SMILES CCCCCC1CCC(CC1)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 170.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Amylcyclohexanol (cis- and trans- mixture)
RTECSNumber GW0680000
TSCA No