missing translation for 'onlineSavingsMsg'
Learn More

4-Amylaniline 97.0+%, TCI America™

Catalog No. A093825ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A093825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A093825ML Supplier TCI America Supplier No. A093825ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33228-44-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00007926
InChI Key DGFTWBUZRHAHTH-UHFFFAOYSA-N
Synonym benzenamine, 4-pentyl, 4-amylaniline, p-pentylaniline, p-amylaniline, unii-8roj0g9hiu, p-n-pentylaniline, 4-n-pentylaniline, 4-pentylphenylamine, 8roj0g9hiu, p-n-amylaniline
PubChem CID 93162
IUPAC Name 4-pentylaniline
SMILES CCCCCC1=CC=C(C=C1)N

Specifications

Boiling Point 265°C
Quantity 25 mL
Formula Weight 163.26
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Amylaniline
EINECSNumber (3)-0206
TSCA Yes