missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminothiomorpholine 1,1-Dioxide 95.0+%, TCI America™
$197.59 - $602.10
Chemical Identifiers
| CAS | 26494-76-8 |
|---|---|
| Molecular Formula | C4H10N2O2S |
| Molecular Weight (g/mol) | 150.196 |
| InChI Key | YDKNASMIEZGJEJ-UHFFFAOYSA-N |
| Synonym | 1,1-Dioxo-4-thiomorpholinamine, 4-Thiomorpholinamine 1,1-Dioxide |
| PubChem CID | 12708263 |
| IUPAC Name | 1,1-dioxo-1,4-thiazinan-4-amine |
| SMILES | C1CS(=O)(=O)CCN1N |
Chemical Identifiers
| 26494-76-8 | |
| 150.196 | |
| 1,1-Dioxo-4-thiomorpholinamine, 4-Thiomorpholinamine 1,1-Dioxide | |
| 1,1-dioxo-1,4-thiazinan-4-amine |
| C4H10N2O2S | |
| YDKNASMIEZGJEJ-UHFFFAOYSA-N | |
| 12708263 | |
| C1CS(=O)(=O)CCN1N |
Specifications
| 26494-76-8 | |
| White-Yellow | |
| 1 g | |
| YDKNASMIEZGJEJ-UHFFFAOYSA-N | |
| 1,1-dioxo-1,4-thiazinan-4-amine | |
| 12708263 | |
| ≥95.0% (GC,T) | |
| 4-Aminothiomorpholine 1,1-Dioxide |
| 107°C | |
| C4H10N2O2S | |
| 1,1-Dioxo-4-thiomorpholinamine, 4-Thiomorpholinamine 1,1-Dioxide | |
| C1CS(=O)(=O)CCN1N | |
| 150.196 | |
| 150.20 | |
| Crystalline Powder |
Safety and Handling
TSCA : No