missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminothiomorpholine 1,1-Dioxide 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 107°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 150.20 |
| Percent Purity | ≥95.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Aminothiomorpholine 1,1-Dioxide |
Safety and Handling
| TSCA | No |