missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyrimidine-5-carbonitrile 98.0+%, TCI America™

Catalog No. A12141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12141G 1 g
A12145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12141G Supplier TCI America Supplier No. A12141G

Chemical Identifiers

CAS 16357-69-0
Molecular Formula C5H4N4
Molecular Weight (g/mol) 120.12
MDL Number MFCD00054552
InChI Key JAIYUIOGVNRXEW-UHFFFAOYSA-N
Synonym 4-amino-5-pyrimidinecarbonitrile, 4-amino-pyrimidine-5-carbonitrile, 4-amino-5-cyanopyrimidine, 5-pyrimidinecarbonitrile, 4-amino, pubchem6999, acmc-1busu, 5-pyrimidinecarbonitrile,4-amino, 4-imino-5-cyano-3,4-dihydropyrimidine, 5-pyrimidinecarbonitrile, 4-amino-6ci,7ci,8ci,9ci
PubChem CID 737208
IUPAC Name 4-aminopyrimidine-5-carbonitrile
SMILES NC1=NC=NC=C1C#N

Specifications

Melting Point 255°C
Quantity 1 g
UN Number 3439
Formula Weight 120.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminopyrimidine-5-carbonitrile