missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyridine 99.0+%, TCI America™

Catalog No. p-7116885
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A041425G 25 g
A0414100G 100 g
A0414500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A041425G Supplier TCI America Supplier No. A041425G
Only null left

Chemical Identifiers

CAS 504-24-5
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006439
InChI Key NUKYPUAOHBNCPY-UHFFFAOYSA-N
Synonym 4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
PubChem CID 1727
ChEBI CHEBI:34385
IUPAC Name pyridin-4-amine
SMILES C1=CN=CC=C1N

Specifications

Melting Point 158°C
Boiling Point 287°C
Quantity 25 g
UN Number 2671
Formula Weight 94.12
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminopyridine
EINECSNumber (5)-0724
RTECSNumber US1750000
TSCA Yes