missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyridine, 99+%

Catalog No. p-7042143
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6147014 25 g
AAJ6147022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6147014 Supplier Thermo Scientific Chemicals Supplier No. J6147014
Only null left

A potassium channel blocker | CAS: 504-24-5 | C5H6N2 | 94.117 g/mol

A potassium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 504-24-5
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00006439
InChI Key NUKYPUAOHBNCPY-UHFFFAOYSA-N
Synonym 4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
PubChem CID 1727
ChEBI CHEBI:34385
IUPAC Name pyridin-4-amine
SMILES C1=CN=CC=C1N

Specifications

Melting Point 157°C to 161°C
Color White
Density 1.26
Boiling Point 273°C to 274°C
Flash Point 164°C (327°F)
Odor Unpleasant
Quantity 25 g
UN Number UN2671
Beilstein 105782
Merck Index 14,3933
Formula Weight 94.12
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminopyridine
Show More Show Less
Hazard Category H300-H311-H315-H319-H335
Hazard Statement GHS H Statement
H300-H311-H315-H319-H335
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES
EINECSNumber 207-987-9
RTECSNumber US1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only