missing translation for 'onlineSavingsMsg'
Learn More

4-Aminopyridine, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 104570010

Catalog No. AC104570010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-Aminopyridine
504-24-5
100.0
156°C
97.5% min. (GC)
C5H6N2
MFCD00006439
22,433
4-aminopyridine, fampridine, 4-pyridinamine, dalfampridine, 4-pyridylamine, avitrol, p-aminopyridine, 4-ap, ampyra, gamma-aminopyridine
NUKYPUAOHBNCPY-UHFFFAOYSA-N
pyridin-4-amine
1727
94.12
98%
97.5
273°C
Authentic
Glass bottle
(C5H4N)NH2
1kg
15,2802
Solubility in water: 74g/L water (20°C). Other solubilities: soluble in ethanol,benzene and ether,slightly soluble in acetone and methanol
C1=CN=CC=C1N
94.12
CHEBI:34385
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Fatal if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-987-9

RTECSNumber : US1750000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only