missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 100047-36-7 |
|---|---|
| Molecular Formula | C6H6N2O2 |
| Molecular Weight (g/mol) | 138.12 |
| InChI Key | JRZBTJVSAANBEV-UHFFFAOYSA-N |
| Synonym | 4-aminopicolinic acid, 4-aminopyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 4-amino, 4-amino-2-pyridinecarboxylic acid, 4-amino-pyridine-2-carboxylic acid, 4-aminopicolinicacid, 4-amino-picolinsaure, pubchem22241, 2-carboxy-4-aminopyridine, picolinic acid, 4-amino |
| PubChem CID | 2050132 |
| IUPAC Name | 4-aminopyridine-2-carboxylic acid |
| SMILES | C1=CN=C(C=C1N)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC455772500
|
N/A | 250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 100047-36-7 | |
| 138.12 | |
| 4-aminopicolinic acid, 4-aminopyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 4-amino, 4-amino-2-pyridinecarboxylic acid, 4-amino-pyridine-2-carboxylic acid, 4-aminopicolinicacid, 4-amino-picolinsaure, pubchem22241, 2-carboxy-4-aminopyridine, picolinic acid, 4-amino | |
| 4-aminopyridine-2-carboxylic acid |
| C6H6N2O2 | |
| JRZBTJVSAANBEV-UHFFFAOYSA-N | |
| 2050132 | |
| C1=CN=C(C=C1N)C(=O)O |
Specifications
| 100047-36-7 | |
| 96% min. (HPLC) | |
| 4-aminopicolinic acid, 4-aminopyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 4-amino, 4-amino-2-pyridinecarboxylic acid, 4-amino-pyridine-2-carboxylic acid, 4-aminopicolinicacid, 4-amino-picolinsaure, pubchem22241, 2-carboxy-4-aminopyridine, picolinic acid, 4-amino | |
| C1=CN=C(C=C1N)C(=O)O | |
| 138.12 | |
| 138.12 | |
| 4-Aminopyridine-2-carboxylic acid |
| Authentic | |
| C6H6N2O2 | |
| JRZBTJVSAANBEV-UHFFFAOYSA-N | |
| 4-aminopyridine-2-carboxylic acid | |
| 2050132 | |
| 97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RUO – Research Use Only