missing translation for 'onlineSavingsMsg'
Learn More

4'-Aminopropiophenone, 98%, Thermo Scientific Chemicals

Catalog No. AC401040100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC401040100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC401040100 Supplier Thermo Scientific Chemicals Supplier No. 401040100

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70-69-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00017113
InChI Key FSWXOANXOQPCFF-UHFFFAOYSA-N
Synonym 4'-aminopropiophenone, p-aminopropiophenone, 1-4-aminophenyl propan-1-one, 1-4-aminophenyl-1-propanone, 1-propanone, 1-4-aminophenyl, 4-aminopropiophenone, propiophenone, 4'-amino, usaf uctl-1856, ethyl p-aminophenyl ketone, paraminopropiophenone
PubChem CID 6270
IUPAC Name 1-(4-aminophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 135.0°C to 140.0°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula NH2C6H4C(O)CH2CH3
Quantity 10 g
Merck Index 15,466
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in ethanol, insoluble in diethyl ether
Formula Weight 149.19
Percent Purity 98%
Chemical Name or Material 4'-Aminopropiophenone
Hazard Category GHS Signal Word: Danger
EINECSNumber 200-742-7

RUO – Research Use Only