missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalimide 98.0+%, TCI America™

Catalog No. p-7117161
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09665G 5 g
A096625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09665G Supplier TCI America Supplier No. A09665G
Only null left

Chemical Identifiers

CAS 3676-85-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00041854
InChI Key PXRKCOCTEMYUEG-UHFFFAOYSA-N
Synonym 4-aminophthalimide, 5-aminoisoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 5-amino, 5-amino-1h-isoindole-1,3 2h-dione, 5-amino-isoindole-1,3-dione, phthalimide, 4-amino, 5-amino-2,3-dihydro-1h-isoindole-1,3-dione, 5-amino-2h-isoindole-1,3-dione, 5-amino-2h-benzo c azolidine-1,3-dione, 5-aminophthalimide
PubChem CID 72915
IUPAC Name 5-amino-2,3-dihydro-1H-isoindole-1,3-dione
SMILES NC1=CC=C2C(=O)NC(=O)C2=C1

Specifications

Quantity 5 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophthalimide
EINECSNumber (5)-3354
TSCA Yes