missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalimide, 97%

Catalog No. p-7023467
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1705106 5 g
AAA1705114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1705106 Supplier Thermo Scientific Chemicals Supplier No. A1705106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3676-85-5 | C8H6N2O2 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3676-85-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00041854
InChI Key PXRKCOCTEMYUEG-UHFFFAOYSA-N
Synonym 4-aminophthalimide, 5-aminoisoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 5-amino, 5-amino-1h-isoindole-1,3 2h-dione, 5-amino-isoindole-1,3-dione, phthalimide, 4-amino, 5-amino-2,3-dihydro-1h-isoindole-1,3-dione, 5-amino-2h-isoindole-1,3-dione, 5-amino-2h-benzo c azolidine-1,3-dione, 5-aminophthalimide
PubChem CID 72915
IUPAC Name 5-aminoisoindole-1,3-dione
SMILES NC1=CC=C2C(=O)NC(=O)C2=C1

Specifications

Melting Point 289°C to 291°C
Quantity 5 g
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material 4-Aminophthalimide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 222-948-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only