missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophthalhydrazide, 98%

Catalog No. AAJ6492103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6492103 1 g
AAJ6492106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6492103 Supplier Thermo Scientific Chemicals Supplier No. J6492103

A chemiluminescent molecule | CAS: 3682-14-2 | C8H7N3O2 | 177.16 g/mol

A chemiluminescent molecule

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3682-14-2
Molecular Formula C8H7N3O2
Molecular Weight (g/mol) 177.16
MDL Number MFCD00010560
InChI Key HUDPLKWXRLNSPC-UHFFFAOYSA-N
Synonym 4-aminophthalhydrazide, isoluminol, 6-aminophthalazine-1,4-diol, chembl14882, 1,4-phthalazinedione, 6-amino-2,3-dihydro, 6-amino-2,3-dihydro-1,4-phthalazinedione, iso-luminol, 1enu, 4-aminophthaloylhydrazine, 4-amino-phthalsaeure-hydrazid
PubChem CID 95014
SMILES NC1=CC=C2C(=O)NNC(=O)C2=C1

Specifications

Melting Point 300°C
Color White to Yellow
Quantity 1 g
Beilstein 164804
Sensitivity Air Sensitive
Solubility Information Soluble in ammonium hydroxide,sodium carbonate or other base.
IUPAC Name 6-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione
Formula Weight 177.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Aminophthalhydrazide
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only