missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylsulfur Pentafluoride 95.0+%, TCI America™

Catalog No. A20435G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A20435G Supplier TCI America Supplier No. A20435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2993-24-0
Molecular Formula C6H6F5NS
Molecular Weight (g/mol) 219.17
MDL Number MFCD01076203
InChI Key MZGZUHNSMNNSRJ-UHFFFAOYSA-N
Synonym 4-aminophenylsulfur pentafluoride, 4-aminophenylsulphur pentafluoride, 1-amino-4-pentafluoro sulfanyl benzene, 4-pentafluorothio aniline, 4-pentafluoro-lambda6-sulfanyl aniline, 4-pentafluoro-??-sulfanyl aniline, acmc-1cjzr, 4-pentafluorosulfanylaniline, 4-pentafluorosulfanyl aniline, 4-pentafluorosulphanyl aniline
PubChem CID 2779190
IUPAC Name 4-(pentafluoro-λ⁶-sulfanyl)aniline
SMILES NC1=CC=C(C=C1)S(F)(F)(F)(F)F

Specifications

Melting Point 64°C
Quantity 5 g
UN Number 2811
Formula Weight 219.17
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophenylsulfur Pentafluoride
TSCA No
Recommended Storage Refrigerator