missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylacetic Acid 98.0+%, TCI America™

Catalog No. A039325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A039325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A039325G Supplier TCI America Supplier No. A039325G

Chemical Identifiers

CAS 1197-55-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007916
InChI Key CSEWAUGPAQPMDC-UHFFFAOYSA-N
Synonym 4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
PubChem CID 14533
IUPAC Name 2-(4-aminophenyl)acetic acid
SMILES NC1=CC=C(CC(O)=O)C=C1

Specifications

Melting Point 192°C
Color Yellow
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminophenylacetic Acid
RTECSNumber AF3550000
TSCA Yes