missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenylacetic acid, 98%

Catalog No. AAA1467530
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1467530 250 g
AAA1467509 10 g
AAA1467518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1467530 Supplier Thermo Scientific Chemicals Supplier No. A1467530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1197-55-3 | C8H9NO2 | 151.17 g/mol

4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminophenylacetic acid, is used as a non-translocated competitive inhibitor of the epithelial peptide transporter PepT1. It can be detected using HPLC, NMR techniques.

Solubility
Very soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents.Stable under normal temperatures and pressures. Stable at room temperature in closed containers under normal storage and handling conditions.

Chemical Identifiers

CAS 1197-55-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007916
InChI Key CSEWAUGPAQPMDC-UHFFFAOYSA-N
Synonym 4-aminophenylacetic acid, 2-4-aminophenyl acetic acid, benzeneacetic acid, 4-amino, p-aminophenylacetic acid, 4-aminobenzeneacetic acid, 4-carboxymethylaniline, acetic acid, p-aminophenyl, para-aminophenyl acetic acid, p-aminophenyl acetic acid, 4-aminophenyl acetic acid
PubChem CID 14533
IUPAC Name 2-(4-aminophenyl)acetic acid
SMILES NC1=CC=C(CC(O)=O)C=C1

Specifications

Melting Point ∼201°C (decomposition)
Beilstein 2208094
Merck Index 14,463
Solubility Information Very soluble in water.
Quantity 250 g
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material 4-Aminophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-828-7
RTECSNumber AF3550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only