missing translation for 'onlineSavingsMsg'
Learn More

4-Aminophenyl phosphate monosodium salt hydrate, ≥98.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111012423
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012423 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012423 Supplier Merck Emd Millipore Supplier No. 75113100MG

Chemical Identifiers

Synonym p-Aminophenyl phosphate monosodium salt hydrate

Specifications

CAS 108084-47-5
Linear Formula C6H7 NNaO4 P · xH2 O
Quantity 100 mg
Formula Weight 211.09 (anhydrous basis)
Percent Purity ≥98.0% (TLC)
Grade For electrochemical analysis