Learn More
4-Aminonicotinic acid, 98%, Thermo Scientific Chemicals
Description
4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine. It is also used as pharmaceutical intermediate.
Solubility
Soluble in aqueous acid and base.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 5 g |
| Solubility Information | Soluble in aqueous acid and base. |
| Formula Weight | 138.13 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Aminonicotinic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only