missing translation for 'onlineSavingsMsg'
Learn More

4-Aminonicotinic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH5003206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5003206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5003206 Supplier Thermo Scientific Chemicals Supplier No. H5003206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7418-65-7 | Molecular Formula: C6H6N2O2 | Molecular Weight: 138.126 g/mol

4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminonicotinic acid has been used in the preparation of 2-methyl-pyrido-oxazine. It is also used as pharmaceutical intermediate.

Solubility
Soluble in aqueous acid and base.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7418-65-7
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00234183
InChI Key IASBMUIXBJNMDW-UHFFFAOYSA-N
Synonym 4-aminonicotinic acid, 4-amino-3-pyridinecarboxylic acid, 4-amino-nicotinic acid, 4-amino-3-carboxypyridine, 4-aminonicotinicacid, 4-amino nicotinic acid, 3-pyridinecarboxylic acid, 4-amino, 4-amino-3-pyridine carboxylic acid, zlchem 370, pubchem1214
PubChem CID 319979
ChEBI CHEBI:68577
IUPAC Name 4-aminopyridine-3-carboxylic acid
SMILES C1=CN=CC(=C1N)C(=O)O

Specifications

Melting Point >300°C
Quantity 5 g
Solubility Information Soluble in aqueous acid and base.
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 4-Aminonicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only