missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomorpholine 98.0+%, TCI America™

Catalog No. p-7117078
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08325ML 5 mL
A083225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08325ML Supplier TCI America Supplier No. A08325ML
Only null left

Chemical Identifiers

CAS 4319-49-7
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD00006174
InChI Key MKQLBNJQQZRQJU-UHFFFAOYSA-N
Synonym 4-aminomorpholine, n-aminomorpholine, 4-morpholinamine, morpholin-4-ylamine, morpholine, 4-amino, 4-morpholinamine 9ci, morpholine-4-ylamine, n-amino morpholine, n-amino-morpholine, 4-amino morpholine
PubChem CID 20315
IUPAC Name morpholin-4-amine
SMILES C1COCCN1N

Specifications

Color Yellow
Boiling Point 168°C
Quantity 5 mL
UN Number 2733
Formula Weight 102.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Aminomorpholine
RTECSNumber QD7300000
TSCA No