missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyltetrahydropyran 98.0+%, TCI America™

Catalog No. A25501G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25501G 1 g
A25505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25501G Supplier TCI America Supplier No. A25501G

Chemical Identifiers

CAS 130290-79-8
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD02179435
InChI Key IPBPLHNLRKRLPJ-UHFFFAOYSA-N
Synonym 4-aminomethyltetrahydropyran, 4-aminomethyl tetrahydro-2h-pyran, tetrahydro-2h-pyran-4-yl methanamine, 4-aminomethyl tetrahydropyran, 1-tetrahydro-2h-pyran-4-ylmethanamine, tetrahydropyran-4-yl methylamine, c-tetrahydro-pyran-4-yl-methylamine, tetrahydro-2h-pyran-4-yl methylamine, 2h-pyran-4-methanamine, tetrahydro, oxan-4-yl methanamine
PubChem CID 2773210
IUPAC Name oxan-4-ylmethanamine
SMILES C1COCCC1CN

Specifications

Color Yellow
Boiling Point 76°C
Quantity 1 g
UN Number 2735
Formula Weight 115.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Aminomethyltetrahydropyran