missing translation for 'onlineSavingsMsg'
Learn More

4-Aminomethyltetrahydropyran, ≥97%, Thermo Scientific™

Catalog No. CC29913CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC29913CB 250mg
CC29913DA 1g
CC29913DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC29913CB Supplier Thermo Scientific Chemicals Supplier No. CC29913CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 130290-79-8
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD02179435
InChI Key IPBPLHNLRKRLPJ-UHFFFAOYSA-N
Synonym 4-aminomethyltetrahydropyran, 4-aminomethyl tetrahydro-2h-pyran, tetrahydro-2h-pyran-4-yl methanamine, 4-aminomethyl tetrahydropyran, 1-tetrahydro-2h-pyran-4-ylmethanamine, tetrahydropyran-4-yl methylamine, c-tetrahydro-pyran-4-yl-methylamine, tetrahydro-2h-pyran-4-yl methylamine, 2h-pyran-4-methanamine, tetrahydro, oxan-4-yl methanamine
PubChem CID 2773210
IUPAC Name oxan-4-ylmethanamine
SMILES C1COCCC1CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 1.02g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material 1-Tetrahydro-2H-pyran-4-ylmethanamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.